Learn More
Medchemexpress LLC 2-methylthio-ATP tetrasodium | 100020-57-3 | 98.0% | 641.20 g/mol | C11H14N5Na4O13P3S | 1 MG

Supplier: Medchemexpress LLC HY1013701MG
2-Methylthio-ATP tetrasodium is the tetrasodium salt of the ATP analogue 2-methylthio-ATP. It is used in pharmacological and receptor-binding studies to activate P2X and P2Y purinergic receptors and to probe platelet aggregation pathways.
- Acts as non-specific P2 receptor agonist.
- Causes noncompetitive inhibition of ADP-induced human platelet aggregation.
- Tetrasodium salt with high water solubility (H2O: 50 mg/mL).
- High purity (98.0%) for consistent experimental results.
- Stable as a sealed solid at -20°C; solutions stable at -80°C up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.