Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M195325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M195325G 25 g
M19535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M195325G Supplier TCI America Supplier No. M195325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3724-10-5
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00029934
InChI Key LWJQGKJCZOGGPJ-UHFFFAOYSA-N
Synonym 2-methylthio benzoic acid, benzoic acid, 2-methylthio, o-methylthio benzoic acid, benzoic acid, o-methylthio, 2-methylsulfanyl benzoic acid, 2-methylmercaptobenzoic acid, 2-carboxyphenyl methyl sulfide, acide methyl-s-2-benzoique, 2-methylmercapto benzoic acid, acide methyl-s-2-benzoique french
PubChem CID 19498
IUPAC Name 2-methylsulfanylbenzoic acid
SMILES CSC1=CC=CC=C1C(=O)O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methylthio)benzoic Acid
RTECSNumber DH4390000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.