Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylthio)phenol, 97+%

Catalog No. AAL1012806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1012806 5 g
AAL1012803 1 g
AAL1012814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1012806 Supplier Thermo Scientific Chemicals Supplier No. L1012806
Add to Cart

CAS: 1073-29-6 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-29-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.20
MDL Number MFCD00002211
InChI Key SOOARYARZPXNAL-UHFFFAOYSA-N
Synonym 2-methylthio phenol, 2-hydroxythioanisole, 2-methylsulfanyl phenol, 2-hydroxy thioanisole, phenol, 2-methylthio, o-hydroxythioanisole, o-methylthio phenol, phenol, o-methylthio, 2-methylmercaptophenol, 2-methylmercapto phenol
PubChem CID 61261
IUPAC Name 2-methylsulfanylphenol
SMILES CSC1=CC=CC=C1O

Specifications

Density 1.16
Boiling Point 104°C to 106°C (22 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.593
Quantity 5 g
Beilstein 1859745
Formula Weight 140.21
Percent Purity ≥97%
Chemical Name or Material 2-(Methylthio)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-027-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.