Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylthiophene, 98%

Catalog No. AAA1273622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1273622 100 g
AAA1273614 25 g
AAA1273636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1273622 Supplier Thermo Scientific Chemicals Supplier No. A1273622
Add to Cart

CAS: 554-14-3 | C5H6S | 98.163 g/mol

2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylthiophene, is used as a flavoring agent that naturally occurs in grilled and roasted beef, papaya, cooked shrimp, whiskey, and in trace amounts in roasted chicken;

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.163
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1

Specifications

Melting Point -63°C
Density 1.017
Boiling Point 112°C to 114°C
Flash Point 7°C (44°F)
Odor Aromatic
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 103734
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 98.17
Percent Purity 98%
Chemical Name or Material 2-Methylthiophene
Show More Show Less
Hazard Category H225-H315-H319-H332-H335
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 209-063-0
RTECSNumber XM9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.