Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltryptamine 98.0+%, TCI America™
SDP

Catalog No. M20551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20551G 1 g
M20555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20551G Supplier TCI America Supplier No. M20551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2731-06-8
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.25
MDL Number MFCD00130185
InChI Key CPVSLHQIPGTMLH-UHFFFAOYSA-N
Synonym 3-(2-Aminoethyl)-2-methylindole
PubChem CID 75949
IUPAC Name 2-(2-methyl-1H-indol-3-yl)ethanamine
SMILES CC1=C(C2=CC=CC=C2N1)CCN

Specifications

Melting Point 108°C
Boiling Point 177°C
Quantity 1 g
Formula Weight 174.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyltryptamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.