Learn More
2-Methylvaleric acid, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
(R,S)-2-methylvaleric acid is an intermediate in the synthesis of branched-chain. 2-Methylpentanoic acid (2-Methylvaleric acid) was used as an internal standard for gas chromatographic analysis of microbial end products.
Solubility
Soluble in water(13g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Specifications
| Melting Point | -85°C |
| Density | 0.93 |
| Boiling Point | 196°C to 197°C |
| Flash Point | 91°C (195°F) |
| Odor | Unpleasant |
| Refractive Index | 1.414 |
| Quantity | 2500 g |
| UN Number | UN3265 |
| Beilstein | 1720655 |
| Solubility Information | Soluble in water(13g/L). |
| Show More |
Safety and Handling
| Hazard Category | H227-H314 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 202-594-9 |
| RTECSNumber | YV7700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.