Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ 2 mL Eye Dropper, Pack of 12
SDPSCI_ED

Catalog No. S81066 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S81066 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81066 Supplier United Scientific™ Supplier No. BPB002PK/12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eye Dropper for use with dropping bottles.

  • Glass pipettes with tapered rubber bulbs
  • For general use or use with dropping bottles
  • Prop 65 Warning Label

Specifications

Material Flint Glass
Sterility Non-sterile
Graduated No
Graduations No
Color Clear
Volume (Metric) 2 mL
Length (Metric) 70 mm
Type Controlled Drop Pasteur Pipet
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.