Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Morpholinomethyl)benzaldehyde 97%, Thermo Scientific™

Catalog No. CC48604CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC48604CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48604CB Supplier Thermo Scientific Chemicals Supplier No. CC48604CB
Add to Cart

Chemical Identifiers

CAS 736991-21-2
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.26
MDL Number MFCD07782412
InChI Key YPZNTOVVELIJGI-UHFFFAOYSA-N
Synonym 2-morpholinomethyl benzaldehyde, 2-morpholin-4-ylmethyl benzaldehyde, morpholinomethylbenzaldehyde, 4-2-carboxaldehydebenzyl morpholine, 2-morpholin-4-yl methyl benzaldehyde
PubChem CID 18525826
IUPAC Name 2-[(morpholin-4-yl)methyl]benzaldehyde
SMILES O=CC1=CC=CC=C1CN1CCOCC1

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 205.26
Chemical Name or Material 2-(morpholinomethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.