Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(n-Butoxycarbonylpiperidine)-5-methyl-1,3-thiazol-4-yl)acetic acid, 97%, Thermo Scientific™

Catalog No. MO07259DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07259DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07259DA Supplier Thermo Scientific Chemicals Supplier No. MO07259DA
Add to Cart

Chemical Identifiers

CAS 845885-88-3
Molecular Formula C16H24N2O4S
Molecular Weight (g/mol) 340.438
InChI Key IHHZQULPYDMAOP-UHFFFAOYSA-N
Synonym 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-thiazol-4-yl acetic acid, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl aceticacid, 2-5-methyl-2-n-tert-butoxycarbonylpiperidine-1,3-thiazol-4-yl acetic acid, 2-5-methyl-2-piperidine-n-boc protected-1,3-thiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-t, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-y, 2-1-butoxycarbonylpiperidine-5-methyl-1,3-thiazol-4-yl acetic acid
PubChem CID 2794810
IUPAC Name 2-[5-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazol-4-yl]acetic acid
SMILES CC1=C(N=C(S1)C2CCN(CC2)C(=O)OC(C)(C)C)CC(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 340.44
Percent Purity 97%
Chemical Name or Material {2-[1-(tert-Butoxycarbonyl)piperidin-4-yl]-5-methyl-1, 3-thiazol-4-yl}acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.