Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Butoxyethyl acetate, 98%

Catalog No. AAL095150F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL095150F 2500 mL
AAL09515AE 100 mL
AAL09515AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL095150F Supplier Thermo Scientific Chemicals Supplier No. L095150F
  • $148.80
    $141.65 / Each of 1

    Save $7.15

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 112-07-2 | C8H16O3 | 160.21 g/mol

2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

Solubility
Miscible with hydrocarbons and organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-07-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00009457
InChI Key NQBXSWAWVZHKBZ-UHFFFAOYSA-N
Synonym butoxyethyl acetate, butyl glycol acetate, butylglycol acetate, ethylene glycol monobutyl ether acetate, 2-butoxyethanol acetate, butyl cellosolve acetate, ektasolve eb acetate, ethanol, 2-butoxy-, acetate, butylcelosolvacetat, 1-acetoxy-2-butoxyethane
PubChem CID 8160
IUPAC Name 2-butoxyethyl acetate
SMILES CCCCOCCOC(C)=O

Specifications

Melting Point -64°C
Density 0.942
Boiling Point 191°C to 192°C
Flash Point 71°C (159°F)
Odor Fruit-like
Refractive Index 1.413
Linear Formula CH3COOCH2CH2O(CH2)3CH3
Quantity 2500 mL
Beilstein 1756960
Solubility Information Miscible with hydrocarbons and organic solvents. Slightly miscible with water.
Formula Weight 160.21
Percent Purity 98%
Chemical Name or Material 2-n-Butoxyethyl acetate
Show More Show Less
Hazard Category H227-H312+H332
Hazard Statement GHS H Statement
H312-H332-H227
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c
EINECSNumber 203-933-3
RTECSNumber KJ8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.