Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-n-Propylheptanoic acid, 97%

Catalog No. AAH5741403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5741403 1 g
AAH57414MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5741403 Supplier Thermo Scientific Chemicals Supplier No. H5741403
Add to Cart

CAS: 31080-39-4 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31080-39-4
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD01723758
InChI Key RXGPYPPCEXISOV-UHFFFAOYNA-N
Synonym 2-propylheptanic acid, 4-nonanecarboxylic acid, heptanoic acid,2-propyl, heptanoic acid, 2-propyl, 2-propylheptansaeure german, 2-n-propylheptanoic acid, 2-n-propyl-1-heptanoic acid, 2-propylheptansaeure, 2-02-00-00313 beilstein handbook reference
PubChem CID 35728
IUPAC Name 2-propylheptanoic acid
SMILES CCCCCC(CCC)C(O)=O

Specifications

Color Colorless to Yellow
Quantity 1 g
Formula Weight 172.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-n-Propylheptanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber MJ2320000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.