Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthol 99.0+%, TCI America™
SDP

Catalog No. N0027250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0027250G 250 g
N002725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0027250G Supplier TCI America Supplier No. N0027250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135-19-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00004067
InChI Key JWAZRIHNYRIHIV-UHFFFAOYSA-N
Synonym 2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
PubChem CID 8663
ChEBI CHEBI:10432
IUPAC Name naphthalen-2-ol
SMILES C1=CC=C2C=C(C=CC2=C1)O

Specifications

Melting Point 121°C
Color White-Yellow
Boiling Point 286°C
Quantity 250 g
Formula Weight 144.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthol
EINECSNumber (4)-0355
RTECSNumber QL2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.