Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthonitrile 98.0+%, TCI America™
SDP

Catalog No. N003925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N003925G 25 g
N00395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N003925G Supplier TCI America Supplier No. N003925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-46-7
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00016807
InChI Key AZKDTTQQTKDXLH-UHFFFAOYSA-N
Synonym 2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
PubChem CID 11944
IUPAC Name naphthalene-2-carbonitrile
SMILES C1=CC=C2C=C(C=CC2=C1)C#N

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 156°C
Quantity 25 g
UN Number 3439
Formula Weight 153.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthonitrile
RTECSNumber QL5976350
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.