Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-2-methyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Catalog No. AC134801000
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC134801000 100 g
AC134800050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC134801000 Supplier Thermo Scientific Chemicals Supplier No. 134801000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-49-6
Molecular Formula C4H9NO4
Molecular Weight (g/mol) 135.12
MDL Number MFCD00024812
InChI Key LOTYADDQWWVBDJ-UHFFFAOYSA-N
Synonym 2-methyl-2-nitro-1,3-propanediol, nmpd, 2-nitro-2-methyl-1,3-propanediol, nitroisobutylglycol, 1,3-propanediol, 2-methyl-2-nitro, 2-methyl-2-nitropropanediol, 1,1-dimethylol-1-nitroethane, caswell no. 573n, epa pesticide chemical code 573900, acmc-1basy
PubChem CID 6480
IUPAC Name 2-methyl-2-nitropropane-1,3-diol
SMILES CC(CO)(CO)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Quantity 100 g
Formula Weight 135.12
Chemical Name or Material 2-Nitro-2-methyl-1, 3-propanediol, 99%
EINECSNumber 201-031-4
RTECSNumber TY8925000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.