Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-4-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. N072025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N072025G 25 g
N07205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N072025G Supplier TCI America Supplier No. N072025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 320-94-5
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
InChI Key MYSAXQPTXWKDPQ-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
PubChem CID 642096
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 235.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.