Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-m-cresol 98.0+%, TCI America™
SDP

Catalog No. N055225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N055225G 25 g
N05525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N055225G Supplier TCI America Supplier No. N055225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4920-77-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007125
InChI Key QIORDSKCCHRSSD-UHFFFAOYSA-N
Synonym 2-nitro-3-methylphenol, 2-nitro-m-cresol, 2-nitro-3-cresol, m-cresol, 2-nitro, phenol, 3-methyl-2-nitro, 2-hydroxy-6-methyl-nitrobenzene, 2-nitro-3-hydroxytoluene, unii-i4qbb2l3c0, 3-methyl-2-nitro-phenol, 3-hydroxy-2-nitrotoluene
PubChem CID 21026
IUPAC Name 3-methyl-2-nitrophenol
SMILES CC1=C(C(=CC=C1)O)[N+](=O)[O-]

Specifications

Melting Point 40°C
Boiling Point 108°C
Quantity 25 g
UN Number 2446
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-m-cresol
EINECSNumber (3)-0790
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.