Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-N-(2-nitrophenyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH6019303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6019303 1 g
AAH60193MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6019303 Supplier Thermo Scientific Chemicals Supplier No. H6019303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331256-09-8
Molecular Formula C12H9N3O6S
Molecular Weight (g/mol) 323.279
MDL Number MFCD00406329
InChI Key NGHQCODQNBAKLD-UHFFFAOYSA-N
Synonym 2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
PubChem CID 2827773
IUPAC Name 2-nitro-N-(2-nitrophenyl)benzenesulfonamide
SMILES C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 323.28
Percent Purity 97%
Chemical Name or Material 2-Nitro-N-(2-nitrophenyl)benzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.