Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzenesulfonyl Chloride 95.0+%, TCI America™
SDP

Catalog No. N0142100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0142100G 100 g
N014225G 25 g
N0142500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0142100G Supplier TCI America Supplier No. N0142100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1694-92-4
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.611
MDL Number MFCD00007430
InChI Key WPHUUIODWRNJLO-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonyl chloride, o-nitrobenzenesulfonyl chloride, o-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, 2-nitro, 2-nitrobenzenesulphonyl chloride, 2-nitrobenzenesulfonylchloride, 2-nitro-benzenesulfonyl chloride, benzenesulfonyl chloride, o-nitro, 2-nitrophenylsulfonyl chloride, benzenesulfonyl chloride, nitro
PubChem CID 15540
IUPAC Name 2-nitrobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)Cl

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 100 g
UN Number 3261
Formula Weight 221.61
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzenesulfonyl Chloride
EINECSNumber (3)-2010
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.