Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzotrifluoride, 98%, Thermo Scientific™

Catalog No. AC166521000
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166521000 100 g
AC166520010 1 g
AC166520050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166521000 Supplier Thermo Scientific Chemicals Supplier No. 166521000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 384-22-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD00007145
InChI Key NDZJSUCUYPZXPR-UHFFFAOYSA-N
Synonym 2-nitrobenzotrifluoride, 1-nitro-2-trifluoromethyl benzene, o-nitrobenzotrifluoride, 2-nitro-alpha,alpha,alpha-trifluorotoluene, o-trifluoromethyl nitrobenzene, benzene, 1-nitro-2-trifluoromethyl, unii-ovb3l8givj, ccris 2813, benzene, nitro trifluoromethyl, ovb3l8givj
PubChem CID 9795
IUPAC Name 1-nitro-2-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 31.0°C to 32.0°C
Boiling Point 216.0°C to 217.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Linear Formula O2NC6H4CF3
Quantity 100 g
Formula Weight 191.11
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2-Nitrobenzotrifluoride
Hazard Category Danger
Hazard Statement GHS H Statement Harmful if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.