Learn More
Sigma Organic Chemistry 2-Nitrophenol | 5G | 88-75-5 | MFCD00011688
Supplier: Sigma Organic Chemistry N197025G

2-Nitrophenol , 5G
About this item:
CAS #: 88-75-5
MDL #: MFCD00011688
Purity: 98%
Chemical Formula: C6H5NO3
Molecular Weight: 139.11
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.