Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitropropane, 96%, Thermo Scientific™

Catalog No. AC157070010
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157070010 1 kg
AC157075000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157070010 Supplier Thermo Scientific Chemicals Supplier No. 157070010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-46-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
MDL Number MFCD00007397
InChI Key FGLBSLMDCBOPQK-UHFFFAOYSA-N
Synonym propane, 2-nitro, isonitropropane, dimethylnitromethane, nitroisopropane, sec-nitropropane, nipar s-20, beta-nitropropane, 2-np, nipar s-20 solvent, nipar s-30 solvent
PubChem CID 398
ChEBI CHEBI:16037
IUPAC Name 2-nitropropane
SMILES CC(C)[N+](=O)[O-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.988g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Refractive Index 1.3930 to 1.3950 (20°C, 589nm)
Linear Formula (CH3)2CHNO2
Quantity 1 kg
Beilstein 01, 116
Fieser 13,200
Merck Index 15, 6715
Formula Weight 89.09
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 2-Nitropropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.