Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Nitropropane 96500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 130265500ML
Encompass
2-Nitropropane can be converted to mutagenic metabolites by human SULT1A1.
Catalog No. 50-289-8078
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.