Learn More
2-Nitroresorcinol, 98%
Description
It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.
Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 81°C to 85°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 2048819 |
| Solubility Information | Slightly soluble in water. Solubility in methanol is almost transparent. |
| Formula Weight | 155.11 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Nitroresorcinol |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 210-010-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.