Learn More
Medchemexpress LLC 2'-O-(2-methoxyethyl)-uridine | 223777-15-9 | 99.2% | 302.28 | C12H18N2O7 | 100 MG

Supplier: Medchemexpress LLC HYW048495100MG
2'-O-(2-methoxyethyl)-uridine is a 2'-modified uridine nucleoside analog used in research. It is a synthetic derivative of uridine employed in studies of nucleoside analogs and has potential application in chemotherapeutic agent development.
- High purity suitable for research use (99.2%).
- Molecular weight 302.28; chemical formula C12H18N2O7.
- Soluble in DMSO (100 mg/mL; may require sonication).
- Storage: solid at 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
- Available as a 100 MG solid quantity and in other sizes for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.