Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-O-Methylguanosine Hydrate 98.0+%, TCI America™
SDP

Catalog No. M23181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23181G 1 g
M2318200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23181G Supplier TCI America Supplier No. M23181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2140-71-8
Molecular Formula C11H15N5O5
Molecular Weight (g/mol) 297.27
MDL Number MFCD00057053
InChI Key OVYNGSFVYRPRCG-YPLCUDRINA-N
Synonym 2'-o-methylguanosine, 2'-o-methyl guanosine, 2'-o-methyl-guanosine, guanosine, 2'-o-methyl, unii-w722h4pa1s, 2-amino-9-2-o-methyl-beta-d-ribofuranosyl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxyoxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-4-hydroxy-5-hydroxymethyl-3-methoxytetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-o-methylguanosine
PubChem CID 188959
ChEBI CHEBI:19229
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxyoxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Specifications

Melting Point 236°C
Color White
Quantity 1 g
Formula Weight 297.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-O-Methylguanosine Hydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.