Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octyl-1-dodecanol, Spectrum™ Chemical
SDP

Catalog No. 18608242 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-242 25 mL Amber Glass Bottle
18-608-244 500 mL Amber Glass Bottle
18-608-243 4 L Amber Glass Bottle
18-608-240 20 L Amber Glass Bottle
18-608-241 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-242 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O201425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5333-42-6

Specifications

CAS Max % 1
Boiling Point 290°C
Packaging Amber Glass Bottle
Quantity 25 mL
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Octyl-1-Dodecanol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.