Learn More
AdipoGen 2-Pefluorohexyl ethyl thiol
SDP
Supplier:  AdipoGen CDXT0478G005
Encompass_Preferred
Chemical. CAS 34451-26-8. Formula C8H5F13S. MW 380.17. Fluorine-containing thiol building block or intermediate for synthesis. Used in fluorofor labeling, surface functionalization of substrates, production of fluorous nanoplatforms and synthesis of biochemical reagents.
Catalog No. 50-255-9940
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.