Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentanone, Spectrum™ Chemical
SDP

Catalog No. 18621240 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-240 100 mL
18-621-241 500 mL
18-621-242 1 L
18-621-243 4 L
18-621-244 4 x 4 L
18-621-245 20 L
18-621-246 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-621-240 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1019100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-87-9

Specifications

Boiling Point 102°C (215.6°F)
Quantity 100 mL
Molecular Weight (g/mol) 86.13
Percent Purity 90%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material 2-Pentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.