Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentene (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. P0064500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0064500ML 500 mL
P006425ML 25 mL
P0064100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0064500ML Supplier TCI America Supplier No. P0064500ML

Chemical Identifiers

CAS 109-68-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
IUPAC Name (2Z)-pent-2-ene
SMILES CC\C=C/C

Specifications

Color Colorless
Boiling Point 36°C
Quantity 500 mL
UN Number 3295
Formula Weight 70.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentene (cis- and trans- mixture)
EINECSNumber (2)-0019
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.