Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. P21211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21211G 1 g
P21215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21211G Supplier TCI America Supplier No. P21211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 721-04-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00156689
InChI Key KRSXGTAVHIDVPM-UHFFFAOYSA-N
PubChem CID 222171
ChEBI CHEBI:52401
IUPAC Name 2-phenoxy-1-phenylethan-1-one
SMILES O=C(COC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 178°C
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.