Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxynicotinic acid, 98%

Catalog No. AAA1124706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1124706 5 g
AAA1124703 1 g
AAA1124714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1124706 Supplier Thermo Scientific Chemicals Supplier No. A1124706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 35620-71-4 | C12H9NO3 | 215.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35620-71-4
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00014629
InChI Key CQGAXJGXGLVFGJ-UHFFFAOYSA-N
Synonym 2-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 2-phenoxy, 2-phenoxy-3-pyridinecarboxylic acid, 2-phenoxy-nicotinic acid, acide 2-phenoxy nicotinique french, acide 2-phenoxy nicotinique, acmc-20a2ql, maybridge1_000744, cambridge id 5832716, 2-phenoxy nicotinic acid
PubChem CID 123450
IUPAC Name 2-phenoxypyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)OC2=C(C=CC=N2)C(=O)O

Specifications

Melting Point 179°C to 182°C
Quantity 5 g
Beilstein 172906
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material 2-Phenoxynicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-647-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.