Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxypropanol 96.0+%, TCI America™
SDP

Catalog No. P111125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111125G 25 g
P11115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P111125G Supplier TCI America Supplier No. P111125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4169-04-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00142958
InChI Key LOJHHQNEBFCTQK-UHFFFAOYNA-N
Synonym Propylene Glycol 2-Monophenyl Ether
PubChem CID 20129
IUPAC Name 2-phenoxypropan-1-ol
SMILES CC(CO)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 244°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxypropanol
EINECSNumber (3)-0560
RTECSNumber UB8886000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.