Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxypropionyl Chloride 98.0+%, TCI America™
SDP

Catalog No. P063825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P063825G 25 g
P0638500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P063825G Supplier TCI America Supplier No. P063825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-35-0
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00018810
InChI Key BDSSZTXPZHIYHM-UHFFFAOYSA-N
Synonym 2-phenoxypropionyl chloride, propanoyl chloride, 2-phenoxy, 2-phenoxypropionic acid chloride, alpha-phenoxypropionyl chloride, 2-phenoxy-propionyl chloride, propanoyl chloride,2-phenoxy, propionyl chloride, 2-phenoxy, .alpha.-phenoxypropionyl chloride, acmc-1bvfg, 2-phenoxypropionylchloride
PubChem CID 95436
IUPAC Name 2-phenoxypropanoyl chloride
SMILES CC(C(=O)Cl)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 g
UN Number 3265
Formula Weight 184.62
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Phenoxypropionyl Chloride
EINECSNumber (3)-2838&(9)-1147&(9)-1258
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.