Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-2-butenal, (E)+(Z), 97%

Catalog No. AAA1418514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418514 25 g
AAA1418506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1418514 Supplier Thermo Scientific Chemicals Supplier No. A1418514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4411-89-6 | C10H10O | 146.19 g/mol

2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in oil and ethanol. Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4411-89-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00053158
InChI Key DYAOGZLLMZQVHY-MBXJOHMKSA-N
Synonym 2-phenyl-2-butenal, 2-phenylcrotonaldehyde, 2-phenylbutenal, alpha-phenylcrotonaldehyde, unii-n8emo6yrv9, e-2-phenylbut-2-enal, 2-butenal, 2-phenyl, alpha-ethylidenebenzeneacetaldehyde, n8emo6yrv9, e-2-phenyl-2-butenal
PubChem CID 6429333
IUPAC Name (E)-2-phenylbut-2-enal
SMILES C\C=C(\C=O)C1=CC=CC=C1

Specifications

Boiling Point 115°C (15 mmHg)
Refractive Index 1.561
Quantity 25 g
Sensitivity Air Sensitive
Solubility Information Soluble in oil and ethanol. Insoluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 2-Phenyl-2-butenal, (E)+(Z)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-567-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.