Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-2-imidazoline, 98%

Catalog No. AAH5909509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5909509 10 g
AAH5909518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5909509 Supplier Thermo Scientific Chemicals Supplier No. H5909509

CAS: 936-49-2 | C9H10N2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005180
InChI Key BKCCAYLNRIRKDJ-UHFFFAOYSA-N
Synonym 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
PubChem CID 13639
IUPAC Name 2-phenyl-4,5-dihydro-1H-imidazole
SMILES C1CN=C(N1)C1=CC=CC=C1

Specifications

Melting Point 99°C to 104°C
Color Brown
Flash Point 201°C (394°F)
Quantity 10 g
Beilstein 119250
Formula Weight 146.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Phenyl-2-imidazoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 213-313-4
RTECSNumber NJ4395500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.