Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. P23841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23841G 1 g
P2384200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23841G Supplier TCI America Supplier No. P23841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 879291-27-7
Molecular Formula C17H20BNO2
Molecular Weight (g/mol) 281.16
MDL Number MFCD12032565
InChI Key XLBHFDXPYKKVFI-UHFFFAOYSA-N
Synonym 4,4,5,5-Tetramethyl-2-(6-phenyl-3-pyridyl)-1,3,2-dioxaborolane, 6-Phenyl-3-pyridylboronic Acid Pinacol Ester
PubChem CID 45786526
IUPAC Name 2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 109°C
Color White
Quantity 1 g
Formula Weight 281.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.