Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylbenzimidazole, 97%

Catalog No. p-7081149
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 716-79-0 | C13H10N2 | 194.24 g/mol

2-Phenylbenzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylbenzimidazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-79-0
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
MDL Number MFCD00005592
InChI Key DWYHDSLIWMUSOO-UHFFFAOYSA-N
Synonym 2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole
PubChem CID 12855
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=C1

Specifications

Melting Point 296°C to 297°C
Quantity 5 g
Beilstein 7087
Merck Index 14,7274
Solubility Information Slightly soluble in water.
IUPAC Name 2-phenyl-1H-1,3-benzodiazole
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 2-Phenylbenzimidazole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-939-2
RTECSNumber DE0360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.