Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylindole-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. P22731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22731G 1 g
P22735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22731G Supplier TCI America Supplier No. P22731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25365-71-3
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00435481
InChI Key IFIFXODAHZPTEY-UHFFFAOYSA-N
Synonym 2-phenylindole-3-carboxaldehyde, 2-phenylindole-3-carbaldehyde, 2-phenyl-3-formylindole, 3-formyl-2-phenylindole, 1h-indole-3-carboxaldehyde, 2-phenyl, 2-phenyl-3-formyl-indole, 3-formyl-2-phenyl indole, cambridge id 5402328, acmc-1cq37, 2-phenyl-3-indolecarboxaldehyde
PubChem CID 613305
IUPAC Name 2-phenyl-1H-indole-3-carbaldehyde
SMILES O=CC1=C(NC2=CC=CC=C12)C1=CC=CC=C1

Specifications

Melting Point 253°C
Quantity 1 g
Formula Weight 221.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylindole-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.