Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. P19451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19451G 1 g
P1945200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19451G Supplier TCI America Supplier No. P19451G

Chemical Identifiers

CAS 7431-45-0
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD04038761
InChI Key OXPDQFOKSZYEMJ-UHFFFAOYSA-N
PubChem CID 593578
IUPAC Name 2-phenylpyrimidine
SMILES C1=CC=C(C=C1)C1=NC=CC=N1

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 156.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Phenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.