Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylsulfonylacetophenone 98.0+%, TCI America™
SDP

Catalog No. P082810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082810G 10 g
P08285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P082810G Supplier TCI America Supplier No. P082810G

Chemical Identifiers

CAS 3406-03-9
Molecular Formula C14H12O3S
Molecular Weight (g/mol) 260.31
MDL Number MFCD00025043
InChI Key DREVPGKOIZVPQV-UHFFFAOYSA-N
Synonym 2-phenylsulfonyl acetophenone, 2-phenylsulfonylacetophenone, 1-phenyl-2-phenylsulfonyl ethanone, acetophenone, 2-phenylsulfonyl, 2-benzenesulfonyl-1-phenylethanone, ethanone, 1-phenyl-2-phenylsulfonyl, 1-phenyl-2-phenylsulphonyl ethan-1-one, alpha-phenylsulfonylacetophenone, 2-benzenesulfonyl-1-phenylethan-1-one, 1-phenyl-2-phenylsulfonyl ethan-1-one
PubChem CID 76949
IUPAC Name 2-(benzenesulfonyl)-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 10 g
Formula Weight 260.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylsulfonylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.