Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylthiazolidine, 97%, Thermo Scientific™

Catalog No. AAH6282303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6282303 1 g
AAH62823MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6282303 Supplier Thermo Scientific Chemicals Supplier No. H6282303

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4569-82-8
Molecular Formula C9H11NS
Molecular Weight (g/mol) 165.254
MDL Number MFCD00128238
InChI Key NXOMVTLTYYYYPC-UHFFFAOYSA-N
Synonym 2-phenylthiazolidine, 2-phenyl-1,3-thiazolane, thiazolidine, 2-phenyl, phenyl-2 thiazolidine, phenyl-2 thiazolidine french, 2-phenyl-thiazolidine, 2-phenyltetrahydrothiazole, phenylthiazolane, phenylthiazolidin, 2-phenyl thiazolidine
PubChem CID 20706
IUPAC Name 2-phenyl-1,3-thiazolidine
SMILES C1CSC(N1)C2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 165.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Phenylthiazolidine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber XJ6094000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.