Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylthiophene 95.0+%, TCI America™
SDP

Catalog No. P13971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13971G 1 g
P13975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13971G Supplier TCI America Supplier No. P13971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 825-55-8
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.234
MDL Number MFCD00130080
InChI Key PJRGDKFLFAYRBV-UHFFFAOYSA-N
Synonym thiophene, 2-phenyl, 2-phenyl-thiophene, phenylthiophene, thiophene, phenyl, 2-thienylbenzene, 2-phenyithiophene, 2-phenylthiofuran, 2-phenylthiophene, acmc-209ppe
PubChem CID 69999
IUPAC Name 2-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=CS2

Specifications

Melting Point 36°C
Color White
Quantity 1 g
Formula Weight 160.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.