Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperazinecarboxylic Acid Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. P139125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P139125G 25 g
P13915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P139125G Supplier TCI America Supplier No. P139125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3022-15-9
Molecular Formula C5H12Cl2N2O2
Molecular Weight (g/mol) 203.063
MDL Number MFCD00044803
InChI Key WNSDZBQLMGKPQS-UHFFFAOYSA-N
Synonym piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
PubChem CID 2723757
IUPAC Name piperazine-2-carboxylic acid;dihydrochloride
SMILES C1CNC(CN1)C(=O)O.Cl.Cl

Specifications

Melting Point 258°C
Quantity 25 g
Formula Weight 203.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Piperazinecarboxylic Acid Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.