Learn More
Medchemexpress LLC 2-piperidinecarboxylic acid ester linked to SLF and thalidomide-based cereblon ligand | 1799711-22-0 | 98.0% | 1009.11 g/mol | C53H64N6O14 | 10 MG

Supplier: Medchemexpress LLC HY10363410MG
dFKBP-1 is a PROTAC small molecule designed to induce degradation of FKBP12 by linking an FKBP12-binding SLF ligand to a thalidomide-derived cereblon ligand via a tailored linker. It is supplied as a characterized research reagent for degrader studies.
- Potent PROTAC degrader of FKBP12 for cellular and biochemical studies.
- Contains an SLF-based FKBP12 ligand and a thalidomide-derived cereblon recruiter.
- High purity (98.0%) with 1H NMR and LCMS confirmation.
- Molecular weight ~1009.11 g/mol; formula C53H64N6O14.
- Off-white to light yellow solid; store at 4°C under inert gas; in solution store at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.