Learn More
Medchemexpress LLC 2-piperidinone, 1-[(2E)-3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propen-1-yl]-3-(trimethoxyphenyl)methylene | 2293951-01-4 | 98.0% | 453.48 g/mol | C25H27NO7 | 10 MG

Supplier: Medchemexpress LLC HY14855310MG
Anti-inflammatory agent 36 is a small-molecule research compound reported to inhibit LPS-induced macrophage activation and to reduce inflammation in preclinical models. It is provided as a high-purity powder for in vitro and in vivo studies, with recommended storage and solvent handling instructions.
- Inhibits LPS-induced macrophage activation in vitro and in vivo.
- High purity (98.0% by HPLC).
- Molecular formula C25H27NO7; molecular weight 453.48 g/mol.
- CAS number 2293951-01-4.
- Soluble in DMSO at 25 mg/mL for cell experiments.
- Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.