Learn More
Eisco™ 2 Prong Chemical Resistant PVC Coated Universal Clamp with Boss head for Large Rods (up 21mm dia.)
SCI_ED
Catalog No. S24284 Shop All Eisco Products
Change view
Click to view available options
Color:
Silver
Catalog No. Color
S24284 Silver
1 options
This item is not returnable. View return policy
Catalog No. S24284 Supplier Eisco™ Supplier No. CH0688D
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
This heavy duty stainless steel clamp comes with chemical resistant PVC coated jaws, each with their own thumbscrew for tightening from both sides. The boss head is able to accommodate up to a 21mm rod and has a thumbscrew for tightening.
Specifications
Type 2-Prong Clamp
Material Zinc Die Cast, Plastic Coated
Color Silver
Jaw Type 2-Prong