Learn More
United Scientific™ 2-Prong Extension Clamp With Steel Rod
SDPSCI_ED
Catalog No. S15909 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
S15909 Black
1 options
Catalog No. S15909 Supplier United Scientific™ Supplier No. CLEX01
Add to Cart
Add to Cart
Sturdy Clamp that can be used with virtually all support stands and lattice setups.
  • 2-prong clamp attached to a chrome-plated steel extension rod.
  • Jaws are lined with cork
  • For use in temperatures up to 60°C
  • Overall length is 8 in.
Specifications
Quantity 1 Ea.
Type Clamp
Material Metal
Color Black
Jaw Type 2-Prong Single Adjust
Warning: Cancer and reproductive harm