Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018964 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8964 5 mL
11-102-0417 1 mL
11-101-8533 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8964 Supplier MilliporeSigma Supelco Supplier No. 912375MLF
Add to Cart
Add to Cart

2-Propanol is a secondary alcohol, which can find a wide number of applications as a solvent.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-63-0
Ignition Point 750°F
Vapor Pressure 44 mmHg (25°C)
Vapor Density 2.1 (vs. air)
Density 0.785 g/mL (at 25°C)
Flash Point 12°C
Physical Form Neat
Linear Formula (CH3)2 CHOH
MDL Number MFCD00011674
Melting Point -89.5°C
Boiling Point 82°C
Synonym sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol
Solubility Completely soluble in water
Type Analytical Standard
Quantity 5 mL
Formula Weight 60.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.