Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ACS, ISO, Reag. Ph Eur., For Analysis, EMSURE™ MilliporeSigma™

Catalog No. M1096344000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
5 L
180 L
Packaging:
Glass Bottle
PET Barrel
Plastic Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1096344000 4 L Glass Bottle
M1096341000 1 L Glass Bottle
M1096341011 1 L Plastic Bottle
M1096342500 2.5 L Glass Bottle
M1096342511 2.5 L Plastic Bottle
M1096345000 5 L Plastic Bottle
M1096349180 180 L PET Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. M1096344000 Supplier MilliporeSigma Supplier No. 1.09634.4000
Add to Cart
Add to Cart
  • Used for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Density 0.786g/cm3
Quantity 4 L
Explosion Limit 2 to 13.4% (V)
Flash Point 12°C
Ignition Point 425°C
Odor Alcoholic
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation <0.001%
Vapor Pressure 43hPa (at 20°C)
Linear Formula C3H8O
UN Number UN 1219
Boiling Point 81°C to 83°C
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity >99.8%
Grade ACS/ISO/Reagent Ph. Eur.
Chemical Name or Material Isopropyl alcohol
Show More Show Less
Hazard Category Irritant
Hazard Statement R 11-36-67: Highly flammable. Irritating to eyes. Vapors may cause drowsiness and dizziness
Precautionary Statement S 7-16-24/25-26: Keep container tightly closed. Keep away from sources of ignition - No smoking. Avoid contact with skin and eyes. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice
EINECSNumber 200-661-7
RTECSNumber NT8050000
Recommended Storage Store at 5° to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.