Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ACS Reagent, ≥99.5%, Solstice

Catalog No. 60045068 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
6 x 1 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-068 1 L Glass Bottle
60-045-071 500 mL Glass Bottle
60-045-069 2.5 L Glass Bottle
60-045-073 6 x 500 mL Glass Bottle
60-045-072 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-068 Supplier Solstice Supplier No. 1907641L
Add to Cart
Add to Cart

ACS reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Density 0.785g/mL at 25°C
Quantity 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.377(lit.)
Vapor Pressure 44mmHg (25°C), 33mmHg (20°C)
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Boiling Point 83°C
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Isopropanol
EINECSNumber 200-661-7
RTECSNumber NT8050000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.